missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (MLANA/7448R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20560-100ug
This item is not returnable.
View return policy
Description
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Melan-A/MART-1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| Recombinant fragment corresponding to the N-terminus of human Melan-A/MART-1 protein | |
| 100 μg | |
| Cell Biology, Cellular Markers, Immunology | |
| 2315 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MLANA/7448R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction