missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (SPM342), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34765H
This item is not returnable.
View return policy
Description
Melan-A/MART-1 Monoclonal specifically detects Melan-A/MART-1 in Human, Equine, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Melan-A/MART-1 | |
| Monoclonal | |
| HRP | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes a protein doublet of 20-22kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This monoclonal antibody labels melanomas and other tumors showing melanocytic differentiation. It is also a useful positive-marker for angiomyolipomas. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. | |
| Store at 4C in the dark. | |
| IgG2b λ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation | |
| SPM342 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MLANA | |
| Recombinant Melan-A/MART-1 protein (Uniprot: Q16655) | |
| 0.1 mL | |
| Cytoskeleton Markers, Immunology | |
| 2315 | |
| Human, Equine, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction