missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (SPM555), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34797CL1
This item is not returnable.
View return policy
Descrizione
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Canine,Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifica
| Melan-A/MART-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| Recombinant Melan-A/MART-1 protein (Uniprot: Q16655) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology | |
| 2315 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| SPM555 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto