missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Melan-A Monoclonal Antibody (CL12864)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562997
This item is not returnable.
View return policy
Description
Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 67%.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
| Melan-A | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16655 | |
| MLANA | |
| Recombinant protein corresponding to Human MLANA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL12864 | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Mouse | |
| Protein A | |
| RUO | |
| 2315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction