missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MelanA (MLANA) Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813599
This item is not returnable.
View return policy
Description
MelanA (MLANA) Monoclonal antibody specifically detects MelanA (MLANA) in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MelanA (MLANA) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P11137, P20357 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 17756, 4133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI1A2 | |
| Unconjugated | |
| Map2 | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Human recombinant protein fragment corresponding to amino acids 1507-1827 of human MAP2 (NP_002365) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction