missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-1 R/MC1R Antibody (SR1585), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Melanocortin-1 R/MC1R |
|---|---|
| Clone | SR1585 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642075
|
Novus Biologicals
NBP3-21979-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653315
|
Novus Biologicals
NBP3-21979-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melanocortin-1 R/MC1R Monoclonal antibody specifically detects Melanocortin-1 R/MC1R in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Melanocortin-1 R/MC1R | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2 | |
| A synthesized peptide derived from human Melanocortin-1 R/MC1R (Uniprot #: Q01726 ) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1585 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4157 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title