missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-3 R/MC3R Antibody (345404), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3737APCCY7
This item is not returnable.
View return policy
Description
Melanocortin-3 R/MC3R Monoclonal antibody specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry
Specifications
| Melanocortin-3 R/MC3R | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 345404 | |
| Flow Cytometry | |
| BMIQ9, MC3, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, OB20, Obesity quantitative trait locus, OQTL | |
| Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R | |
| 0.1 mL | |
| GPCR | |
| 4159 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction