missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-4 R Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41103AF750
This item is not returnable.
View return policy
Description
Melanocortin-4 R Polyclonal antibody specifically detects Melanocortin-4 R in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Melanocortin-4 R | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MC4-R, melanocortin 4 receptor, melanocortin receptor 4, MGC126851, MGC138197 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MC4R. The immunogen is located within the first 50 amino acids of MC4R. Amino Acid Squence: SYRLHSNASESLGKGYSDG | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 4160 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction