missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Associated Antigen (PNL2) Antibody (PNL2), Alexa Fluor™ 350 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11561AF350
This item is not returnable.
View return policy
Description
Melanoma Associated Antigen (PNL2) Monoclonal antibody specifically detects Melanoma Associated Antigen (PNL2) in Human, Mouse, Canine samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Melanoma Associated Antigen (PNL2) | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse, Canine | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PNL2 | |
| 50mM Sodium Borate | |
| Melanocyte antigen | |
| 0.1 mL | |
| Primary | |
| Store at 4&drg;C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction