missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), Novus Biologicals™
Mouse Monoclonal Antibody
€ 274.00 - € 527.00
Specifications
| Antigen | Melanoma Marker (MART-1 + gp100) |
|---|---|
| Clone | DT101+ BC199 + HMB45 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18446101
|
Novus Biologicals
NBP2-34341-20ug |
20 μg |
€ 274.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18427211
|
Novus Biologicals
NBP2-34341-100ug |
100 μg |
€ 527.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Melanoma Marker (MART-1 + gp100) | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16655, Q16655, P40967, Q16655 | |
| 2315 | |
| IgG2b κ, IgG1 κ | |
| Protein A or G purified |
| DT101+ BC199 + HMB45 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| MART1, MART-1 | |
| Recombinant Melanoma Marker (MART-1 + gp100) protein (DT101 & BC199), Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title