missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanophilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17274
This item is not returnable.
View return policy
Description
Melanophilin Polyclonal specifically detects Melanophilin in Mouse samples. It is validated for Western Blot.
Specifications
| Melanophilin | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| exophilin-3, l(1)-3Rk, l1Rk3, ln, melanophilin, MGC2771, MGC59733, Slac-2a, SLAC2A, SLAC2-A, Slp homolog lacking C2 domains a, Synaptotagmin-like protein 2a | |
| Rabbit | |
| 65 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse. Species reactivity based on sequence identity: Rat (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.98mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q91V27 | |
| MLPH | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse Melanophilin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79083 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction