missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanophilin Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079083-B01P
This item is not returnable.
View return policy
Description
Melanophilin Polyclonal antibody specifically detects Melanophilin in Human samples. It is validated for Western Blot
Specifications
| Melanophilin | |
| Polyclonal | |
| PBS (pH 7.4) | |
| exophilin-3, l(1)-3Rk, l1Rk3, ln, melanophilin, MGC2771, MGC59733, Slac-2a, SLAC2A, SLAC2-A, Slp homolog lacking C2 domains a, Synaptotagmin-like protein 2a | |
| MLPH (NP_077006.1, 1 a.a. - 600 a.a.) full-length human protein. MGKKLDLSKLTDEEAQHVLEVVQRDFDLRRKEEERLEALKGKIKKESSKRELLSDTAHLNETHCARCLQPYQLLVNSKRQCLECGLFTCKSCGRVHPEEQGWICDPCHLARVVKIGSLEWYYEHVKARFKRFGSAKVIRSLHGRLQGGAGPELISEERSGDSDQTDEDGEPGSEAQAQAQPFGSKKKRLLSVHDFDFEGDSDDSTQPQGHSLHLSSVPEARDSPQSLTDESCSEKAAPHKAEGLEEADTGASGCHSHPEEQPTSISPSRHGALAELCPPGGSHRMALGTAAALGSNVIRNEQLPLQYLADVDTSDEESIRAHVMASHHSKRRGRASSESQIFELNKHISAVECLLTYLENTVVPPLAKGLGAGVRTEADVEEEALRRKLEELTSNVSDQETSSEEEEAKDEKAEPNRDKSVGPLPQADPEVGTAAHQTNRQEKSPQDPGDPVQYNRTTDEELSELEDRVAVTASEVQQAESEVSDIESRIAALRAAGLTVKPSGKPRRKSNLPIFLPRVAGKLGKRPEDPNADPSSEAKAMAVPYLLRRKFSNSLKSQGKDDDSFDRKSVYRGSLTQRNPNARKGMASHTFAKPVVAHQS | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_077006.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 79083 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction