missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanophilin Antibody (OTI6E8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72649APC
This item is not returnable.
View return policy
Description
Melanophilin Monoclonal antibody specifically detects Melanophilin in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Melanophilin | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79083 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI6E8 | |
| Western Blot, Immunohistochemistry | |
| exophilin-3, l(1)-3Rk, l1Rk3, ln, melanophilin, MGC2771, MGC59733, Slac-2a, SLAC2A, SLAC2-A, Slp homolog lacking C2 domains a, Synaptotagmin-like protein 2a | |
| Human recombinant protein fragment corresponding to amino acids 295-600 of human MLPH(NP_077006) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction