missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanotransferrin/CD228/MFI2 Antibody (893416), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB81751MFV500
This item is not returnable.
View return policy
Description
Melanotransferrin/CD228/MFI2 Monoclonal antibody specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Melanotransferrin/CD228/MFI2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 893416 | |
| Flow Cytometry, CyTOF-ready | |
| antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MGC4856, MTF1 | |
| HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228, Gly20-Gly711, Accession # P08582 | |
| RUO | |
| 4241 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction