Learn More
Novus Biologicals™ Melanotransferrin/CD228/MFI2 Antibody (SC-005) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28837-50ug
 View more versions of this product
Product Code. 30046142
  • € 213.00 / 50µg
Description
Melanotransferrin/CD228/MFI2 Monoclonal antibody specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Melanotransferrin/CD228/MFI2
Monoclonal
Unconjugated
P08582
Human
Protein A purified
RUO
4241
Human
Purified
Flow Cytometry, ELISA, Functional Assay
SC-005
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MGC4856, MTF1
Melanotransferrin/ CD228
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.