missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melatonin R1A/MT1/MTNR1A Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-28912
This item is not returnable.
View return policy
Description
Melatonin R1A/MT1/MTNR1A Polyclonal specifically detects Melatonin R1A/MT1/MTNR1A in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Melatonin R1A/MT1/MTNR1A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Mel1a receptor, MEL-1A-R, melatonin receptor 1A, melatonin receptor type 1A, MT1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4543 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| P48039 | |
| MTNR1A | |
| Peptide with sequence C-SLKYDKLYSSKNS corresponding to internal region according to NP_005949.1. | |
| 0.1 mg | |
| Primary | |
| Expected cross-reactivity from sequence similarity: Mouse, Dog, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction