Learn More
B Braun™ Meliseptol™ Rapid 250 mL Spray Bottle
Product Code. 15981290
Click to view available options
Volume (Metric):
250 mL
Unit Size:
250mL
Product Code. 15981290
Brand: B Braun™ 18564
A fast-acting surface disinfectant spray for resistant surfaces.
  • Ready-to-use alcohol-based disinfectant
  • Completely aldehyde-free and alkylamine-free
  • For surfaces of non-invasive medical devices such as treatment chairs, work areas and operating room equipment
  • Broad efficacy spectrum – bactericidal (including MRSA), tuberculocidal and fungicidal (including molds)
  • Active against enveloped viruses (including HBV, HIV, HCV) and non-enveloped viruses (including Norovirus)
  • Exposure time of only 1 minute for enveloped viruses, 10 minutes for non-enveloped viruses
  • Fresh smell
Specifications
Product Type Disinfectant Spray
pH ∼7
Appearance Colorless
Composition Alcohol-based
Container Type Spray Bottle
For Use With (Application) Rapid disinfection
Volume (Metric) 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Meliseptol rapid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : propan-1-ol, Didecyldimethylammoniumchloride