Learn More
B Braun™ Surface Spray Disinfectant
Product Code. 10381722
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10381722 1 L 1 L
10709372 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10381722 Supplier B Braun™ Supplier No. 18565
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
Use B Braun™ Surface Spray Disinfectant for rapid surface disinfection with Rapid Meliseptrol. The broad-spectrum disinfectant has a 5-minute reaction time.
  • Rapid Meliseptrol distribution
  • Free of CFC's
  • For small surfaces and equipment
  • Colorless
Order Info
VolumeMetric
Specifications
Product Type Disinfectant
Form Liquid
Volume (Metric) 1 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Meliseptol rapid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Propan-1-ol, Didecyldimethylammoniumchlorid