missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melphalan-modified DNA Antibody (MP5/73), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50504B
This item is not returnable.
View return policy
Description
Melphalan-modified DNA Monoclonal specifically detects Melphalan-modified DNA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| Melphalan | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Native DNA reacted with melphalan | |
| 0.1 mL | |
| Cancer | |
| Other | |
| Purified |
| ELISA, Immunohistochemistry | |
| MP5/73 | |
| ELISA, Immunohistochemistry | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction