missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Membralin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17276
This item is not returnable.
View return policy
Description
Membralin Polyclonal specifically detects Membralin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Membralin | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| TMEM259 | |
| Recombinant protein encompassing a sequence within the center region of human Membralin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 91304 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ASBABP1, aspecific BCL2 ARE-binding protein 1, chromosome 19 open reading frame 6, MBRL, membralin, MGC4022, R32184_3 | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction