missing translation for 'onlineSavingsMsg'
Learn More

Membrin Antibody (8G6) [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30520258 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520258 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520258 Supplier Novus Biologicals Supplier No. NBP342976G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Membrin Monoclonal antibody specifically detects Membrin in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Membrin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8G6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias golgi SNAP receptor complex member 2, GS27Bos1,27 kDa Golgi SNARE protein, membrin
Host Species Mouse
Immunogen golgi SNAP receptor complex member 2 - (Uniprot# O14653)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 9570
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.