missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14604-1-AP-150UL
This item is not returnable.
View return policy
Description
May control cell migration by relaying extracellular chemotactic signals to the microtubule cytoskeleton. Mediator of ERBB2 signaling. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization. Is required for breast carcinoma cell migration.Specifications
| MEMO1 | |
| Polyclonal | |
| Unconjugated | |
| MEMO1 | |
| C21orf19 like protein, C2orf4, CGI 27, mediator of cell motility 1, MEMO, Memo 1, MEMO1, NS5ATP7, Protein MEMO1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298787, 51072, 76890 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4QQR9, Q91VH6, Q9Y316 | |
| MEMO1 | |
| MEMO1 Fusion Protein Ag6151 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction