Learn More
Alexandra™ Men's Lab Coat with Concealed Stud Front, Light Blue
Product Code. p-7145437
Click to view available options
Size (Metric) Chest:
116 cm
124 cm
Unit Size:
Each
Product Code. 15872194
Brand: Alexandra™ WL1PB116
Only null left
Add to basket
Add to basket
Men's stud coat featuring three practical pockets with concealed stud front for a professional look. Durable polyester/cotton construction. D piece in neck can be used to display a bar code to simplify the laundering process.
  • Concealed stud front
  • Two hip pockets one chest pocket
  • Back vent
  • Machine washable and industrially launderable
  • Length 100cm / 39 in.
  • Fabric Weight: 245gsm (210gsm white)
  • 67% polyester/33% cotton
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Men's
Material Polycotton
Color Light Blue
Disposable Reusable
Sterility Non-sterile
Model WL1
Size (English) Chest 46 in.
Size (Metric) Chest 116 cm
Weight (Metric) 245 gsm