Learn More
MEN1 Mouse anti-Human, Clone: 7D3E10, Abnova™
Mouse monoclonal antibody raised against partial recombinant human MEN1.
Brand: Abnova MAB12246.100ug
Description
This gene encodes menin, a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Specifications
| MEN1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| MEAI/SCG2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
| 7D3E10 | |
| Mouse monoclonal antibody raised against partial recombinant human MEN1. | |
| In PBS (0.05% sodium azide) | |
| MEN1 | |
| Recombinant protein corresponding to amino acids 392-554 of human MEN1. | |
| RUO | |
| 4221 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.