missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mena Antibody (ENAH/1988), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79887APC
This item is not returnable.
View return policy
Description
Mena Monoclonal antibody specifically detects Mena in Human samples. It is validated for Western Blot, ELISA, Protein Array
Specifications
| Mena | |
| Monoclonal | |
| APC | |
| ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog | |
| Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) (Uniprot: Q8N8S7) | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 55740 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Western Blot, ELISA, Protein Array | |
| ENAH/1988 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction