missing translation for 'onlineSavingsMsg'
Learn More

Menadione, MedChemExpress

Product Code. 16442106
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442106 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442106 Supplier Medchem Express Supplier No. HYB0332
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Menadione, a synthetic naphthoquinone, can be converted to active vitamin K2 in vivo.

Menadione, a synthetic naphthoquinone, can be converted to active vitamin K2 in vivo.

Specifications

Chemical Name or Material Menadione
CAS 58-27-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8O2
Quantity 500 mg
Synonym Vitamin K3
Solubility Information DMSO : 50 mg/mL (290.39 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(C)=CC(C2=C1C=CC=C2)=O
Molecular Weight (g/mol) 172.18
Formula Weight 172.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.