missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menin Antibody, Alexa Fluor™ 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-287AF405
This item is not returnable.
View return policy
Description
Menin Polyclonal antibody specifically detects Menin in Human samples. It is validated for Western Blot
Specifications
| Menin | |
| Polyclonal | |
| Western Blot | |
| menin, multiple, multiple endocrine neoplasia I | |
| The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221). | |
| 0.1 mL | |
| Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
| 4221 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction