Learn More
Menin Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-215R
This item is not returnable.
View return policy
Description
Menin Polyclonal antibody specifically detects Menin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Menin | |
| DyLight 550 | |
| Immunogen affinity purified | |
| Human |
| Immunohistochemistry | |
| The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221). | |
| RUO | |
| IgG |