missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menin Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-287
This item is not returnable.
View return policy
Description
Menin Polyclonal specifically detects Menin in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Menin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| menin, multiple, multiple endocrine neoplasia I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for human Menin. Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Dusky titi monkey, Crab-eating macaque, Olive baboon and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
| NP_000235.2 | |
| MEN1 | |
| The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221). | |
| 100 μL | |
| Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
| 4221 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction