missing translation for 'onlineSavingsMsg'
Learn More

Menin-MLL inhibitor MI-2, MedChemExpress

Product Code. 16401176
Encompass
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16401176

Brand: Medchem Express HY15222

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM.

Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM.

Specifications

Chemical Name or Material Menin-MLL inhibitor MI-2
Physical Form Solid
CAS 1271738-62-5
Color Light Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C18H25N5S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (133.14 mM; Need ultrasonic)
SMILES CCCC1=CC2=C(N3CCN(C4=NCC(C)(C)S4)CC3)N=CN=C2S1
Molecular Weight (g/mol) 375.55
Formula Weight 375.55
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.