missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEOX2 Monoclonal Antibody (OTI4F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805639
This item is not returnable.
View return policy
Description
Mesodermal transcription factor that plays a key role in somitogenesis and is required for sclerotome development. Activates expression of CDKN1A and CDKN2A in endothelial cells, acting as a regulator of vascular cell proliferation. While it activates CDKN1A in a DNA-dependent manner, it activates CDKN2A in a DNA-independent manner. May have a regulatory role when quiescent vascular smooth muscle cells reenter the cell cycle.Specifications
| MEOX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50222 | |
| MEOX2 | |
| Full length human recombit protein of human MEOX2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4F6 | |
| Unconjugated | |
| MEOX2 | |
| GAX, MOX2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4223 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction