missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEOX2 Monoclonal Antibody (OTI4F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805639S
This item is not returnable.
View return policy
Description
MEOX2 Monoclonal antibody specifically detects MEOX2 in Human samples. It is validated for Western BlotSpecifications
| MEOX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75459 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8712 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4F6 | |
| Unconjugated | |
| PAGE1 | |
| AI528662; GAX; Growth arrest-specific homeobox; homeobox protein MOX-2; MEOX2; mesenchyme homeo box 2 (growth arrest-specific homeo box); mesenchyme homeobox 2; Mox2; Mox-2 | |
| Full length human recombinant protein of human PAGE1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction