missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR3 Mouse anti-Human, Clone: OTI6C1, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811551
This item is not returnable.
View return policy
Description
Folate is an essential vitamin that must be obtained from food intake through intestinal absorption in mammals. Folate and reduced folic acid derivatives bind to the folate receptor (FR) family, which mediates the endocytosis of 5-methyltetrahydrofolate into the cell. The folate receptors consist of five members, alpha, beta, gamma, gamma′ (which is produced by alternative splicing) and delta. beta-FR is attached to the membrane by a GPI anchor and is expressed in maligt tissues of epithelial and nonepithelial origin. gamma-FR is expressed in tissues of hematopoietic origin, such as spleen, thymus and bone marrow. The expression pattern of delta-FR is elusive, which suggests that it is highly restricted both spatially and temporally. The alpha isoform of the folate receptor is used as a highly selective tumor marker and may be targeted for the delivery of therapeutic compounds to tumor cells by coupling to derivatives of folic acid.Specifications
| FOLR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41439 | |
| FOLR3 | |
| Full length human recombit protein of human FOLR3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C1 | |
| Unconjugated | |
| FOLR3 | |
| FR-G, FR-gamma, gamma-hFR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction