missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEP1B Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547474
This item is not returnable.
View return policy
Description
In Western blots, approximately 40% cross-reactivity with recombinant human MEP1B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Meprins are multidomain zinc metalloproteases that are highly expressed in mammalian kidney and intestinal brush border membranes, and in leukocytes and certain cancer cells. They are involved in the hydrolysis of a variety of peptide and protein substrates, and have been implicated in cancer and intestinal inflammation. Mature meprins are oligomers of evolutionarily related, but separately encoded alpha and/or beta subunits. Homooligomers of alpha subunit are secreted, whereas, oligomers containing the beta subunit are plasma membrane-bound. This gene encodes the beta subunit. Targeted disruption of this gene in mice affects embryonic viability, renal gene expression profiles, and distribution of the membrane-associated alpha subunit in kidney and intestine.
Specifications
| MEP1B | |
| Polyclonal | |
| Unconjugated | |
| Mep1b | |
| EC 3.4.24.63; Endopeptidase-2; MEP1B; Mep-1b; meprin 1 beta; meprin A; meprin A subunit beta; meprin A, beta; meprin B; meprin beta; N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta; PABA peptide hydrolase; PPH beta | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17288 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61847 | |
| Mep1b | |
| Mouse myeloma cell line NS0-derived recombinant mouse Meprin beta Subunit/MEP1B Leu21-Ser594 (Thr75Ile, Ile432Val). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction