Learn More
Invitrogen™ MEPE Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA547460
Product Code. 15547952
  • € 462.00 / 100µg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MEPE
Polyclonal
Unconjugated
MEPE
Matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; osteoblast/osteocyte factor 45; Osteoregulin
Goat
Antigen affinity chromatography
RUO
56955
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q9NQ76
MEPE
Mouse myeloma cell line NS0-derived recombinant human MEPE/OF45 Pro18-Asp525.