missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEPE Polyclonal Antibody
GREENER_CHOICE

Product Code. 15796081 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796081 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796081 Supplier Invitrogen™ Supplier No. PA558206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: IPASMNYAKA HSKDKKKPQR DSQAQKSPVK SKSTHRIQHN IDYLKHLSKV KKIPSDFEGS GYTDLQERGD NDISPFSGDG QPFKDIPGKG EATGPD Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 48%.

Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEPE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEPE
Gene Accession No. Q9NQ76
Gene Alias Matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; osteoblast/osteocyte factor 45; Osteoregulin
Gene Symbols MEPE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEPE. Recombinant protein control fragment (Product #RP-96693).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.