missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEPE Polyclonal Antibody
GREENER_CHOICE

Product Code. 17732634
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17732634 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17732634 Supplier Invitrogen™ Supplier No. PA5121235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, Mouse liver, Mouse spinal cord, Rat brain, Rat liver. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: FTNRQRLNKE YSISNKENTH NGLRMSIYPK STGNKGFEDG DDAISKLHDQ EEYGAALIRN NMQHIMGPVT AIKLLGEENK ENTPRNVLNI IPASMNYAKA HSKDKKKPQR DSQAQKSPVK SKSTHRIQHN IDYLKHLSKV KKIPSDFEGS GYTDLQERGD NDISPFS.

Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEPE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MEPE
Gene Accession No. D6C6P2, Q8K4L6, Q9NQ76
Gene Alias Matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; osteoblast/osteocyte factor 45; Osteoregulin
Gene Symbols MEPE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-256 of human MEPE (NP_064588.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56955, 79110, 94111
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.