missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meprin alpha Subunit/MEP1A Antibody (364312) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3220I
This item is not returnable.
View return policy
Description
Meprin alpha Subunit/MEP1A Monoclonal antibody specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Meprin alpha Subunit/MEP1A | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF | |
| 364312 | |
| Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
| Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A, Val22-Gln601, Accession # AAA21338 | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 4224 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction