Learn More
Mer Antibody (125518), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8912CL1
Product Code. 30148771
  • ONLINE EXCLUSIVE PRICE
    € 1150.00
    € 1087.80 / 0.10mL
    Save € 62.20
    5% Off
Description
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF,Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mer
Monoclonal
CoraFluor 1
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer, Met1-Ala499, Accession # AAB60430
0.1 mL
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision
10461
Store at 4°C in the dark. Do not freeze.
IgG2b
Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
125518
PBS
Mouse
Protein A or G purified