missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MERS Coronavirus NP Recombinant Rabbit Monoclonal Antibody (1R8L7)
GREENER_CHOICE

Product Code. 18076361
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076361 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18076361 Supplier Invitrogen™ Supplier No. MA551380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MASPAAPRAV SFADNNDITN TNLSRGRGRN PKPRAAPNNT VSWYTGLTQH GKVPLTFPPG QGVPLNANST PAQNAGYWRR QDRKINTGNG IKQLAPRWYF YYTGTGPEAA LPFRAVKDGI VWVHEDGATD APSTFGTRNP NNDSAIVTQF APGTKLPKNF HIEGTGGNSQ SSSRASSLSR NSSRSSSQGS RSGNSTRGTS PGPSGIGAVG GDLLYLDLLN RLQALESGKV KQSQPKVITK KDAAAAKNKM RHKRTSTKSF NMVQAFGLRG PGDLQGNFGD LQLNKLGTED PRWPQIAELA PTASAFMGMS QFKLTHQNND DHGNPVYFLR YSGAIKLDPK NPNYNKWLEL LEQNIDAYKT FPKKEKKQKA PKEESTDQMS EPPKEQRVQG SITQRTRTRP SVQPGPMIDV NTD.

Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MERS Coronavirus NP
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1R8L7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias Coronavirus; MERS; NP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-413 of coronavirus Nucleoprotein (YP_009047211.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.