missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike Protein Antibody (12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 160.00 - € 462.00
Specifications
| Antigen | MERS-CoV Spike Protein |
|---|---|
| Clone | 12 |
| Dilution | Western Blot 1:2000-1:10000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026566
|
Novus Biologicals
NBP3-26065-20ul |
20 μL |
€ 160.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039211
|
Novus Biologicals
NBP3-26065-100ul |
100 μL |
€ 462.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MERS-CoV Spike Protein Monoclonal antibody specifically detects MERS-CoV Spike Protein in Virus samples. It is validated for Western Blot, ELISASpecifications
| MERS-CoV Spike Protein | |
| Western Blot 1:2000-1:10000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
| Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant MERS-CoV Spike Protein (Accession#: AFS88936.1, Met1-Trp1297) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Infections (Virus Bacteria and Parasites) | |
| 0.2 um filtered solution in PBS | |
| 14254594 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title