missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike Protein Antibody - N-terminal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 160.00 - € 459.00
Specifications
| Antigen | MERS-CoV Spike Protein |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, ELISA : 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641584
|
Novus Biologicals
NBP3-20217-20ul |
20 μg |
€ 160.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687234
|
Novus Biologicals
NBP3-20217-100ul |
100 μg |
€ 459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MERS-CoV Spike Protein Polyclonal antibody specifically detects MERS-CoV Spike Protein in MERS-CoV samples. It is validated for Western Blot, ELISASpecifications
| MERS-CoV Spike Protein | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| 0.2 um filtered solution in PBS | |
| 14254594 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000, ELISA : 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| MERS-CoV | |
| MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
| Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the S1 subunit of MERS-CoV Spike Protein | |
| Primary | |
| Store at 4&drg;C short term. Aliquot and store at -20&drg;C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title