missing translation for 'onlineSavingsMsg'
Learn More

MES, >98%, MP Biomedicals™

Product Code. 11445262 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
1 kg
Unit Size:
100g
10g
1kg
250g
25g

Product Code. 11445262

Brand: MP Biomedicals 0219530910

Please to purchase this item. Need a web account? Register with us today!

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-[N-Morpholino]ethanesulfonic acid
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range >98%
Formula Weight 213.2
Chemical Name or Material MES
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.