missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (420411) [DyLight 350], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB32652D
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF, Gene Knock-Out
Specifications
| Mesothelin | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, CyTOF, Gene Knock-Out | |
| 420411 | |
| Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| Mouse myeloma cell line NS0-derived recombinant human Mesothelin, Glu296-Gly580, Accession # AAH09272 | |
| 0.1 mL | |
| Cancer | |
| 10232 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction