missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (CL13956) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24589AF647
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Mesothelin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| This antibody was developed using a recombinant protein derived from Q13421, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 10232 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13956 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction