missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Mesothelin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL13965 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a recombinant protein sequence of Q13421, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?