missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (MB-G10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-85538C
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page
Specifications
| Mesothelin | |
| Monoclonal | |
| DyLight 650 | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| This Mesothelin Antibody (MB-G10) was produced in mesothelin-deficient mice by immunizations with plasmid cDNA encoding human Mesothelin full length protein followed by a single boost of a recombinant human mesothelin-Fc fusion protein. (Uniprot: Q13421) | |
| 0.1 mL | |
| Cancer | |
| 10232 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MB-G10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction