missing translation for 'onlineSavingsMsg'
Learn More

Mesothelin Antibody (MSLN/6443R), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20893DL594

Product Code. 18535138

  • € 762.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mesothelin Monoclonal antibody specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mesothelin
Monoclonal
DyLight 594
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Recombinant fragment (around aa 273-407) of human Mesothelin protein (exact sequence is proprietary)
0.1 mL
Cancer
10232
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
MSLN/6443R
50 mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.