missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MESP2 Antibody (1C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00145873-M01
This item is not returnable.
View return policy
Description
MESP2 Monoclonal antibody specifically detects MESP2 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| MESP2 | |
| Monoclonal | |
| Unconjugated | |
| XP_085261 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 145873 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 1C8 | |
| In 1x PBS, pH 7.4 | |
| bHLHc6, class C basic helix-loop-helix protein 6, mesoderm posterior 2 homolog (mouse), mesoderm posterior protein 2, SCDO2 | |
| MESP2 (XP_085261, 2 a.a. ∽ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AQSPPPQSLLGHDHWIFAQGWGWAGHWDSTSPASSSDSSGSCPCDGARGLPQPQPPSCSSRAAEAAATTPRRARTGPAGGQRQSASEREKLRMRTLARALHELRRFLPPSLAPAGQSLTKIETL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction