missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MESP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17911031
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911031 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911031 Supplier Invitrogen™ Supplier No. PA5143961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human A549 whole cell, human SW620 whole cell. IHC: human lung cancer tissue. ICC/IF: A549 cell. Flow: K562 cell, SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MESP2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene Mesp2
Gene Accession No. Q0VG99
Gene Alias bHLHc6; Class C basic helix-loop-helix protein 6; mesoderm posterior 2; mesoderm posterior 2 homolog; mesoderm posterior basic helix-loop-helix transcription factor 2; mesoderm posterior bHLH transcription factor 2; mesoderm posterior protein 2; Mesp2; SCDO2
Gene Symbols Mesp2
Host Species Rabbit
Immunogen E.coli-derived human Mesp2 recombinant protein (Position: M1-Y397).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145873
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.